(2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetic acid ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A174928-100MG A174928-250MG A174928-1G
77683-405EA 21 USD
77683-405 77683-406 77885-361
(2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetic acid ≥98%, Ambeed.Inc
(2S)-2-Amino-2-(3-hydroxyadamantan-1-yl)acetic acid

Precaución: For research use only.

Formula: C₁₂H₁₉NO₃
MDL Number: MFCD16660446
CAS Number: 709031-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR