(T-4)-Potassium (E)-trifluoro(oct-1-en-1-yl)borate ≥95%

Supplier: Ambeed
Warning

A1743060-250MG A1743060-5G A1743060-1G
77877-247EA 83 USD
77877-247 77877-248 77877-246
(T-4)-Potassium (E)-trifluoro(oct-1-en-1-yl)borate ≥95%
(T-4)-Potassium (E)-trifluoro(oct-1-en-1-yl)borate

Precaución: for Research Use Only

Formula: C₈H₁₅BF₃K
CAS Number: 1227512-01-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR