tert-Butyl (3-(chlorosulfonyl)propyl)carbamate ≥97%, Ambeed.Inc

Supplier: Ambeed
Danger

A174133-100MG A174133-250MG
77683-399EA 238 USD
77683-399 77890-120
tert-Butyl (3-(chlorosulfonyl)propyl)carbamate ≥97%, Ambeed.Inc
tert-Butyl (3-(chlorosulfonyl)propyl)carbamate

Precaución: For research use only.

Formula: C₈H₁₆ClNO₄S
MW: 257.73 g/mol
MDL Number: MFCD13190123
CAS Number: 1196153-59-9
UN: 2928
ADR: 6.1, 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR