(Z)-1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanone O-(2,4-dichlorobenzyl) oxime nitrate ≥98%

Supplier: Ambeed
Danger

Synonyms: Oxiconazole nitrate

A173419-1G A173419-250MG
77671-462EA 815 USD
77671-462 77671-464
(Z)-1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanone O-(2,4-dichlorobenzyl) oxime nitrate ≥98%
(Z)-1-(2,4-Dichlorophenyl)-2-(1H-imidazol-1-yl)ethanone O-(2,4-dichlorobenzyl) oxime nitrate

Precaución: for Research Use Only

Formula: C₁₈H₁₄Cl₄N₄O₄
MDL Number: MFCD01727545
CAS Number: 64211-46-7
UN: 1479
ADR: 5.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR