tert-Butyl (1S,2S,5R)-2-(hydroxymethyl)-3,8-diazabicyclo[3.2.1]octane-8-carboxylate ≥98%
Supplier: Ambeed
|
Warning
|
A1729335-100MG
A1729335-250MG
77625-812EA
103
USD
77625-812
77625-814
tert-Butyl (1S,2S,5R)-2-(hydroxymethyl)-3,8-diazabicyclo[3.2.1]octane-8-carboxylate ≥98%
tert-Butyl (1S,2S,5R)-2-(hydroxymethyl)-3,8-diazabicyclo[3.2.1]octane-8-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₂₂N₂O₃ MW: 242.31 g/mol |
CAS Number:
2820536-71-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...