Flavopiridol ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Chlorophenyl)-5,7-dihydroxy-8-((3S,4R)-3-hydroxy-1-methylpiperidin-4-yl)-4H-chromen-4-one

A172904-1MG A172904-5MG A172904-10MG
77600-398EA 24.6 USD
77600-398 77600-400 77667-410
Flavopiridol ≥98%
Flavopiridol

Precaución: for Research Use Only

Formula: C₂₁H₂₀ClNO₅
MW: 401.840 g/mol
MDL Number: MFCD20501884
CAS Number: 146426-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR