Lithium triisopropoxy(isoquinolin-1-yl)borate ≥95%

Supplier: Ambeed
Danger

A1728028-1G A1728028-250MG A1728028-5G
77877-203EA 199 USD
77877-203 77877-204 77877-205
Lithium triisopropoxy(isoquinolin-1-yl)borate ≥95%
Lithium triisopropoxy(isoquinolin-1-yl)borate

Precaución: for Research Use Only

Formula: C₁₈H₂₇BLiNO₃
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR