(4-(4-(Methoxycarbonyl)quinolin-2-yl)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1727222-100MG A1727222-1G A1727222-250MG
77877-192EA 47 USD
77877-192 77877-193 77877-194
(4-(4-(Methoxycarbonyl)quinolin-2-yl)phenyl)boronic acid ≥95%
(4-(4-(Methoxycarbonyl)quinolin-2-yl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₄BNO₄
MW: 307.11 g/mol
CAS Number: 2102513-94-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR