Methyl 4-(((tert-butoxycarbonyl)amino)methyl)benzoate ≥97%
Supplier: Ambeed
|
Warning
|
A172295-25G
A172295-5G
77890-108EA
595
USD
77890-108
77890-109
Methyl 4-(((tert-butoxycarbonyl)amino)methyl)benzoate ≥97%
Methyl 4-(((tert-butoxycarbonyl)amino)methyl)benzoate
Precaución: for Research Use Only
Formula:
C₁₄H₁₉NO₄ |
MDL Number:
MFCD11870086 CAS Number: 120157-96-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...