2-(tert-Butyl)phenyl trifluoromethanesulfonate ≥98%

Supplier: Ambeed
Danger

A1722085-1G A1722085-250MG A1722085-5G
77877-179EA 217 USD
77877-179 77877-180 77877-181
2-(tert-Butyl)phenyl trifluoromethanesulfonate ≥98%
2-(tert-Butyl)phenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₁₁H₁₃F₃O₃S
CAS Number: 198206-03-0
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR