tert-Butyl (20-bromo-3,6,9,12,15,18-hexaoxaicosyl)carbamate ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Br-PEG6-C2-NHBoc
A1722040-250MG
A1722040-100MG
77600-392EA
155.8
USD
77600-392
77625-796
tert-Butyl (20-bromo-3,6,9,12,15,18-hexaoxaicosyl)carbamate ≥95%
tert-Butyl (20-bromo-3,6,9,12,15,18-hexaoxaicosyl)carbamate
Precaución: for Research Use Only
Formula:
C₁₉H₃₈BrNO₈ MW: 488.41 g/mol |
CAS Number:
2688072-13-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...