MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥99%

Supplier: Ambeed
Warning

Synonyms: (6-Methoxyquinolinio)acetic acid ethyl ester bromide, 1-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide, 1-(2-Ethoxy-2-oxoethyl)-6-methoxyquinolin-1-ium bromide, MQAE

A172174-100MG A172174-250MG A172174-1G
77600-388EA 88.15 USD
77600-388 77600-390 77667-398
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) ≥99%
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide)

Precaución: for Research Use Only

Formula: C₁₄H₁₆NO₃Br
MDL Number: MFCD00467854
CAS Number: 162558-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR