(4-Chloro-1-methyl-1H-indol-2-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1721567-100MG A1721567-1G A1721567-250MG
77876-768EA 47 USD
77876-768 77876-769 77876-770
(4-Chloro-1-methyl-1H-indol-2-yl)boronic acid ≥95%
(4-Chloro-1-methyl-1H-indol-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₉H₉BClNO₂
MW: 209.44 g/mol
MDL Number: MFCD18427579
CAS Number: 912331-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR