(6-Methylpyrimidin-4-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1721561-100MG A1721561-250MG
77876-766EA 209 USD
77876-766 77876-767
(6-Methylpyrimidin-4-yl)boronic acid ≥95%
(6-Methylpyrimidin-4-yl)boronic acid

Precaución: for Research Use Only

Formula: C₅H₇BN₂O₂
MDL Number: MFCD13659297
CAS Number: 1883429-66-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR