(3aR,3a'R,8aS,8a'S)-2,2'-(1,3-Bis(3,5-di-tert-butylphenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) ≥95%
Supplier: Ambeed
|
Warning
|
A1721307-100MG
A1721307-250MG
77598-214EA
107.63
USD
77598-214
77598-216
(3aR,3a'R,8aS,8a'S)-2,2'-(1,3-Bis(3,5-di-tert-butylphenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole) ≥95%
(3aR,3a'R,8aS,8a'S)-2,2'-(1,3-Bis(3,5-di-tert-butylphenyl)propane-2,2-diyl)bis(3a,8a-dihydro-8H-indeno[1,2-d]oxazole)
Precaución: for Research Use Only
Formula:
C₅₁H₆₂N₂O₂ MW: 735.05 g/mol |
CAS Number:
2387715-06-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...