N-((2,4-Dimethylphenyl)sulfonyl)-2-(naphthalen-2-yloxy)acetamide ≥98%

Supplier: Ambeed
Warning

A1719720-10MG A1719720-5MG A1719720-50MG A1719720-100MG A1719720-250MG A1719720-25MG
77625-780EA 108.15 USD
77625-780 77625-782 77877-147 77877-144 77877-145 77877-146
N-((2,4-Dimethylphenyl)sulfonyl)-2-(naphthalen-2-yloxy)acetamide ≥98%
N-((2,4-Dimethylphenyl)sulfonyl)-2-(naphthalen-2-yloxy)acetamide

Precaución: for Research Use Only

Formula: C₂₀H₁₉NO₄S
CAS Number: 868145-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR