Lithium (6-bromopyridin-2-yl)triisopropoxyborate ≥95%

Supplier: Ambeed
Danger

A1719179-1G A1719179-250MG A1719179-5G
77877-138EA 197 USD
77877-138 77877-139 77877-140
Lithium (6-bromopyridin-2-yl)triisopropoxyborate ≥95%
Lithium (6-bromopyridin-2-yl)triisopropoxyborate

Precaución: for Research Use Only

Formula: C₁₄H₂₄BBrLiNO₃
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR