(6-((tert-Butoxycarbonyl)amino)naphthalen-2-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1717725-1G A1717725-5G
77877-132EA 207 USD
77877-132 77877-133
(6-((tert-Butoxycarbonyl)amino)naphthalen-2-yl)boronic acid ≥95%
(6-((tert-Butoxycarbonyl)amino)naphthalen-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₈BNO₄
MDL Number: MFCD24470899
CAS Number: 1131395-21-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR