(4S,4'S)-2,2'-(4-(Trifluoromethyl)pyridine-2,6-diyl)bis(4-methyl-4,5-dihydrooxazole) ≥97%

Supplier: Ambeed
Warning

A1716905-50MG A1716905-100MG
77625-764EA 211.15 USD
77625-764 77877-125
(4S,4'S)-2,2'-(4-(Trifluoromethyl)pyridine-2,6-diyl)bis(4-methyl-4,5-dihydrooxazole) ≥97%
(4S,4'S)-2,2'-(4-(Trifluoromethyl)pyridine-2,6-diyl)bis(4-methyl-4,5-dihydrooxazole)

Precaución: for Research Use Only

Formula: C₁₄H₁₄F₃N₃O₂
CAS Number: 2866174-39-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR