2-(2,6-dioxopiperidin-3-yl)-4-(prop-2-yn-1-yloxy)isoindoline-1,3-dione ≥98%

Supplier: Ambeed
Warning

Synonyms: Thalidomide-propargyl

A171175-100MG A171175-250MG A171175-50MG
77748-131EA 161 USD
77748-131 77748-132 77748-133
2-(2,6-dioxopiperidin-3-yl)-4-(prop-2-yn-1-yloxy)isoindoline-1,3-dione ≥98%
2-(2,6-dioxopiperidin-3-yl)-4-(prop-2-yn-1-yloxy)isoindoline-1,3-dione

Precaución: for Research Use Only

Formula: C₁₆H₁₂N₂O₅
MW: 312.28 g/mol
MDL Number: MFCD32063450
CAS Number: 2098487-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR