2-(6-bromopyridin-2-yl)acetonitrile ≥98%

Supplier: Ambeed
Warning

A170648-1G A170648-250MG
77671-422EA 87 USD
77671-422 77883-319
2-(6-bromopyridin-2-yl)acetonitrile ≥98%
2-(6-bromopyridin-2-yl)acetonitrile

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MDL Number: MFCD08706174
CAS Number: 112575-11-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR