5-Bromo-2-(m-tolyl)-2,3-dihydroquinazolin-4(1H)-one ≥98%

Supplier: Ambeed
Warning

A1705477-10MG A1705477-1MG A1705477-5MG
77598-206EA 120.95 USD
77598-206 77598-208 77598-210
5-Bromo-2-(m-tolyl)-2,3-dihydroquinazolin-4(1H)-one ≥98%
5-Bromo-2-(m-tolyl)-2,3-dihydroquinazolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₁₅H₁₃BrN₂O
CAS Number: 2819989-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR