N-(3-(1H-Imidazol-1-yl)propyl)-5,7-diphenylpyrazolo[1,5-a]pyrimidine-2-carboxamide ≥98%

Supplier: Ambeed
Warning

Synonyms: JNJ-9350

A1702949-5MG A1702949-10MG A1702949-1MG
77598-204EA 138.38 USD
77598-204 77598-200 77598-202
N-(3-(1H-Imidazol-1-yl)propyl)-5,7-diphenylpyrazolo[1,5-a]pyrimidine-2-carboxamide ≥98%
N-(3-(1H-Imidazol-1-yl)propyl)-5,7-diphenylpyrazolo[1,5-a]pyrimidine-2-carboxamide

Precaución: for Research Use Only

Formula: C₂₅H₂₂N₆O
MW: 422.48 g/mol
MDL Number: MFCD01050984
CAS Number: 326923-09-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR