(S)-4-(3-(4-(4-Acryloylpiperazin-1-yl)-6-((S)-2,4-dimethylpiperazin-1-yl)pyridin-2-yl)-1,2,4-oxadiazol-5-yl)-2-amino-4-methyl-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile ≥98%

Supplier: Ambeed
Warning

Synonyms: BI-0474

A1702925-10MG A1702925-1MG A1702925-5MG
77598-194EA 123 USD
77598-194 77598-196 77598-198
(S)-4-(3-(4-(4-Acryloylpiperazin-1-yl)-6-((S)-2,4-dimethylpiperazin-1-yl)pyridin-2-yl)-1,2,4-oxadiazol-5-yl)-2-amino-4-methyl-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile ≥98%
(S)-4-(3-(4-(4-Acryloylpiperazin-1-yl)-6-((S)-2,4-dimethylpiperazin-1-yl)pyridin-2-yl)-1,2,4-oxadiazol-5-yl)-2-amino-4-methyl-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carbonitrile

Precaución: for Research Use Only

Formula: C₃₀H₃₇N₉O₂S
MDL Number: MFCD34789532
CAS Number: 2750570-55-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR