3-Bromo-1-(4-methoxybenzyl)piperidine-2,6-dione ≥95%

Supplier: Ambeed
Danger

A1702335-1G A1702335-25G
77876-715EA 142 USD
77876-715 77876-716
3-Bromo-1-(4-methoxybenzyl)piperidine-2,6-dione ≥95%
3-Bromo-1-(4-methoxybenzyl)piperidine-2,6-dione

Precaución: for Research Use Only

Formula: C₁₃H₁₄BrNO₃
CAS Number: 2300100-12-1
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR