3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine ≥98%

Supplier: Ambeed
Warning

A169971-1G A169971-100G A169971-10G A169971-25G
77671-406EA 16.6 USD
77671-406 77881-308 77881-309 77881-310
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine ≥98%
3-(Trifluoromethyl)-5,6,7,8-tetrahydro[1,2,4]triazolo[4,3-a]pyrazine

Precaución: for Research Use Only

Formula: C₆H₇F₃N₄
MDL Number: MFCD08460920
CAS Number: 486460-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR