5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol ≥98%

Supplier: Ambeed
Warning

A1699420-250MG A1699420-100MG
77625-734EA 239.99 USD
77625-734 77876-712
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol ≥98%
5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol

Precaución: for Research Use Only

Formula: C₁₆H₁₉BO₃
CAS Number: 2323577-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR