Methyl 2-amino-6-methoxybenzoate ≥97%

Supplier: Ambeed
Warning

A169780-100MG A169780-1G A169780-250MG
77883-312EA 18 USD
77883-312 77883-313 77883-314
Methyl 2-amino-6-methoxybenzoate ≥97%
Methyl 2-amino-6-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.19 g/mol
MDL Number: MFCD16036887
CAS Number: 54166-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR