Madecassoside ≥98%

Supplier: Ambeed
Warning

Synonyms: (1S,2R,4aS,6aS,6bR,8R,8aR,9R,10R,11R,12aR,12bR,14bS)-(2S,3R,4S,5S,6R)-6-((((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2S,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)methyl)-3,4,5-trihydroxytetrahydro-2H-pyran-2-yl 8,10,11-trihydroxy-9-(hydroxymethyl)-1,2,6a,6b,9,12a-hexamethyl-1,2,3,4,4a,5,6,6a,6b,7,8,8a,9,10,11,12,12a,12b,13,14b-icosahydropicene-4a-carboxylate

A169218-250MG A169218-100MG
77685-372EA 110 USD
77685-372 77685-371
Madecassoside ≥98%
Madecassoside

Precaución: for Research Use Only

Formula: C₄₈H₇₈O₂₀
MW: 975.13 g/mol
MDL Number: MFCD00017380
CAS Number: 34540-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR