4-Acetamido-3-methylbenzoic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

A169017-1G A169017-10G
77683-383EA 14 USD
77683-383 77890-092
4-Acetamido-3-methylbenzoic acid ≥95%, Ambeed.Inc
4-Acetamido-3-methylbenzoic acid

Precaución: For research use only.

Formula: C₁₀H₁₁NO₃
MDL Number: MFCD08691999
CAS Number: 37901-92-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR