Quinoxalin-6-ylmethanamine hydrochloride ≥95%

Supplier: Ambeed
Warning

A168811-1G A168811-100MG
77611-642EA 201.88 USD
77611-642 77683-380
Quinoxalin-6-ylmethanamine hydrochloride ≥95%
Quinoxalin-6-ylmethanamine hydrochloride

Precaución: For research use only.

Formula: C₉H₉N₃·ClH
MW: 195.649 g/mol
MDL Number: MFCD15424795
CAS Number: 1276056-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR