6-Chloro-N4-(3,5-difluoro-4-((3-methyl-1H-pyrrolo[2,3-b]pyridin-4-yl)oxy)phenyl)pyrimidine-2,4-diamine ≥98%

Supplier: Ambeed
Warning

Synonyms: BAY-549

A168750-1MG A168750-5MG
77615-404EA 45.32 USD
77615-404 77746-130
6-Chloro-N4-(3,5-difluoro-4-((3-methyl-1H-pyrrolo[2,3-b]pyridin-4-yl)oxy)phenyl)pyrimidine-2,4-diamine ≥98%
6-Chloro-N4-(3,5-difluoro-4-((3-methyl-1H-pyrrolo[2,3-b]pyridin-4-yl)oxy)phenyl)pyrimidine-2,4-diamine

Precaución: for Research Use Only

Formula: C₁₈H₁₃ClF₂N₆O
MW: 402.79 g/mol
MDL Number: MFCD15528941
CAS Number: 867017-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR