5-Acetamidopyridine-3-boronic acid pinacol ester ≥95%

Supplier: Ambeed
Warning

A168490-100MG A168490-250MG
77600-350EA 51.25 USD
77600-350 77667-350
5-Acetamidopyridine-3-boronic acid pinacol ester ≥95%
5-Acetamidopyridine-3-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₉BN₂O₃
MDL Number: MFCD11878288
CAS Number: 1201645-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR