(11bS)-2,6-Dibromo-N,N-dimethyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥95%
Supplier: Ambeed
|
Warning
|
A1684571-1G
A1684571-100MG
77625-720EA
1093.86
USD
77625-720
77625-718
(11bS)-2,6-Dibromo-N,N-dimethyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine ≥95%
(11bS)-2,6-Dibromo-N,N-dimethyldinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine
Precaución: for Research Use Only
Formula:
C₂₂H₁₆Br₂NO₂P MW: 517.15 g/mol |
CAS Number:
284472-86-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...