2-Phenyl-1,10-phenanthroline ≥97%

Supplier: Ambeed
Warning

A1684507-250MG A1684507-1G A1684507-25G A1684507-5G
77593-036EA 111.73 USD
77593-036 77598-188 77876-700 77876-701
2-Phenyl-1,10-phenanthroline ≥97%
2-Phenyl-1,10-phenanthroline

Precaución: for Research Use Only

Formula: C₁₈H₁₂N₂
MDL Number: MFCD31793607
CAS Number: 109559-47-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR