Sodium 1-(octadec-9-enoyloxy)-2,5-dioxopyrrolidine-3-sulfonate ≥98%

Supplier: Ambeed
Warning

Synonyms: sulphosuccinimidyl oleate sodium

A1684359-10MG A1684359-1MG A1684359-5MG
77598-182EA 78.93 USD
77598-182 77598-184 77598-186
Sodium 1-(octadec-9-enoyloxy)-2,5-dioxopyrrolidine-3-sulfonate ≥98%
Sodium 1-(octadec-9-enoyloxy)-2,5-dioxopyrrolidine-3-sulfonate

Precaución: for Research Use Only

Formula: C₂₂H₃₆NNaO₇S
MW: 481.58 g/mol
CAS Number: 1212012-37-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR