4-((7-Aminoheptyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%

Supplier: Ambeed
Warning

Synonyms: Pomalidomide-C7-NH2 hydrochloride

A1684215-100MG A1684215-250MG A1684215-50MG A1684215-1G
77598-178EA 152.73 USD
77598-178 77598-180 77625-712 77876-692
4-((7-Aminoheptyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride ≥95%
4-((7-Aminoheptyl)amino)-2-(2,6-dioxopiperidin-3-yl)isoindoline-1,3-dione hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₇ClN₄O₄
MW: 422.91 g/mol
CAS Number: 2446474-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR