Sodium tetrahydrofuran-3-sulfinate ≥98%

Supplier: Ambeed
Warning

A1683355-100MG A1683355-250MG A1683355-1G
77593-034EA 79.95 USD
77593-034 77625-710 77876-685
Sodium tetrahydrofuran-3-sulfinate ≥98%
Sodium tetrahydrofuran-3-sulfinate

Precaución: for Research Use Only

Formula: C₄H₇NaO₃S
MDL Number: MFCD28124335
CAS Number: 1622013-59-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR