2-Amino-4-bromo-5-chloro-3,6-difluorobenzoic acid ≥95%

Supplier: Ambeed
Warning

A1683210-1G A1683210-250MG A1683210-100MG
77598-174EA 290.08 USD
77598-174 77598-176 77625-708
2-Amino-4-bromo-5-chloro-3,6-difluorobenzoic acid ≥95%
2-Amino-4-bromo-5-chloro-3,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrClF₂NO₂
MW: 286.46 g/mol
CAS Number: 2387928-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR