(11aR)-1,10-Bis(3,5-di-tert-butylphenyl)-12-hydroxy-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocine 12-oxide ≥98%

Supplier: Ambeed
Warning

A1682540-100MG A1682540-1G A1682540-250MG
77625-698EA 185.4 USD
77625-698 77625-700 77625-702
(11aR)-1,10-Bis(3,5-di-tert-butylphenyl)-12-hydroxy-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocine 12-oxide ≥98%
(11aR)-1,10-Bis(3,5-di-tert-butylphenyl)-12-hydroxy-4,5,6,7-tetrahydrodiindeno[7,1-de:1',7'-fg][1,3,2]dioxaphosphocine 12-oxide

Precaución: for Research Use Only

Formula: C₄₅H₅₅O₄P
MW: 690.89 g/mol
CAS Number: 1639448-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR