(2-((1,3-Dioxoisoindolin-2-yl)methyl)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1677309-250MG A1677309-100MG A1677309-1G
77876-662EA 83 USD
77876-662 77876-660 77876-661
(2-((1,3-Dioxoisoindolin-2-yl)methyl)phenyl)boronic acid ≥95%
(2-((1,3-Dioxoisoindolin-2-yl)methyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₂BNO₄
CAS Number: 428820-97-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR