(3-((tert-Butyldimethylsilyl)oxy)-5-(methoxycarbonyl)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1677304-1G A1677304-250MG A1677304-5G
77876-651EA 211 USD
77876-651 77876-652 77876-653
(3-((tert-Butyldimethylsilyl)oxy)-5-(methoxycarbonyl)phenyl)boronic acid ≥95%
(3-((tert-Butyldimethylsilyl)oxy)-5-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₃BO₅Si
MW: 310.23 g/mol
MDL Number: MFCD28166642
CAS Number: 1416251-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR