(3,5-Dichloro-4-(methoxycarbonyl)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1677303-250MG A1677303-5G
77876-649EA 81 USD
77876-649 77876-650
(3,5-Dichloro-4-(methoxycarbonyl)phenyl)boronic acid ≥95%
(3,5-Dichloro-4-(methoxycarbonyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₇BCl₂O₄
MW: 248.86 g/mol
MDL Number: MFCD23380395
CAS Number: 1321613-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR