(7-Chloro-1H-indol-2-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1677297-100MG A1677297-1G A1677297-250MG
77876-643EA 45 USD
77876-643 77876-644 77876-645
(7-Chloro-1H-indol-2-yl)boronic acid ≥95%
(7-Chloro-1H-indol-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₇BClNO₂
MW: 195.41 g/mol
MDL Number: MFCD28662105
CAS Number: 1286777-12-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR