(Z)-(2-Bromo-2-phenylvinyl)boronic acid ≥95%

Supplier: Ambeed
Danger

A1676829-1G A1676829-250MG A1676829-5G
77876-592EA 272 USD
77876-592 77876-593 77876-594
(Z)-(2-Bromo-2-phenylvinyl)boronic acid ≥95%
(Z)-(2-Bromo-2-phenylvinyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₈BBrO₂
MW: 226.86 g/mol
CAS Number: 128458-97-9
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR