(3-(3-(3-Chlorophenyl)ureido)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A1676804-5G A1676804-1G A1676804-250MG
77876-582EA 773 USD
77876-582 77876-580 77876-581
(3-(3-(3-Chlorophenyl)ureido)phenyl)boronic acid ≥95%
(3-(3-(3-Chlorophenyl)ureido)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClN₂O₃
MW: 290.51 g/mol
CAS Number: 1400223-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR