(5-Bromo-4-methylthiazol-2-yl)methanamine ≥95%

Supplier: Ambeed
Warning

A167430-1G A167430-250MG
77683-370EA 556 USD
77683-370 77683-371
(5-Bromo-4-methylthiazol-2-yl)methanamine ≥95%
(5-Bromo-4-methylthiazol-2-yl)methanamine

Precaución: For research use only.

Formula: C₅H₇BrN₂S
MW: 207.09 g/mol
MDL Number: MFCD26613560
CAS Number: 1515474-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR