N-((4,6-Dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-5-(ethyl(tetrahydro-2H-pyran-4-yl)amino)-4-methyl-4'-(piperazin-1-ylmethyl)-[1,1'-biphenyl]-3-carboxamide ≥98%

Supplier: Ambeed
Warning

Synonyms: EZH2-IN-13

A1673640-1MG A1673640-5MG A1673640-50MG A1673640-10MG A1673640-25MG
77598-156EA 105.58 USD
77598-156 77625-668 77876-562 77876-560 77876-561
N-((4,6-Dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-5-(ethyl(tetrahydro-2H-pyran-4-yl)amino)-4-methyl-4'-(piperazin-1-ylmethyl)-[1,1'-biphenyl]-3-carboxamide ≥98%
N-((4,6-Dimethyl-2-oxo-1,2-dihydropyridin-3-yl)methyl)-5-(ethyl(tetrahydro-2H-pyran-4-yl)amino)-4-methyl-4'-(piperazin-1-ylmethyl)-[1,1'-biphenyl]-3-carboxamide

Precaución: for Research Use Only

Formula: C₃₄H₄₅N₅O₃
MDL Number: MFCD34676843
CAS Number: 1403255-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR