(R)-2-(2-(2-((3,3-Dibutyl-7-(methylthio)-1,1-dioxido-5-phenyl-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-8-yl)oxy)acetamido)-2-phenylacetamido)acetic acid hydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: Elobixibat hydrate

A1673454-1MG A1673454-100MG A1673454-10MG A1673454-1G A1673454-250MG A1673454-25MG A1673454-50MG
77625-666EA 9.27 USD
77625-666 77876-554 77876-555 77876-556 77876-557 77876-558 77876-559
(R)-2-(2-(2-((3,3-Dibutyl-7-(methylthio)-1,1-dioxido-5-phenyl-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-8-yl)oxy)acetamido)-2-phenylacetamido)acetic acid hydrate ≥98%
(R)-2-(2-(2-((3,3-Dibutyl-7-(methylthio)-1,1-dioxido-5-phenyl-2,3,4,5-tetrahydrobenzo[b][1,4]thiazepin-8-yl)oxy)acetamido)-2-phenylacetamido)acetic acid hydrate

Precaución: for Research Use Only

Formula: C₃₆H₄₇N₃O₈S₂
MW: 713.9 g/mol
CAS Number: 1633824-78-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR