1-(3-Chloropyridin-4-yl)ethanol ≥95%

Supplier: Ambeed
Warning

A167341-1G A167341-250MG
77615-390EA 369.77 USD
77615-390 77748-110
1-(3-Chloropyridin-4-yl)ethanol ≥95%
1-(3-Chloropyridin-4-yl)ethanol

Precaución: for Research Use Only

Formula: C₇H₈ClNO
MDL Number: MFCD24501840
CAS Number: 886856-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR